missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human RECK Fc Chimera Protein

Product Code. 18738236
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18738236

Brand: R&D Systems™ 10309RE050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Human RECK Fc Chimera is immobilized at 2 μg/mL (100 μL/well), Recombinant Human TEM5/GPR124 His-tag binds with an ED50 of 0.7 to 5.6 μg/mL
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95980
For Use With (Application) Bioactivity
Gene ID (Entrez) 8434
Molecular Weight (g/mol) 127 kDa
Name hRECK, membrane-anchored glycoprotein (metastasis and invasion), reversion-inducing-cysteine-rich protein with kazal motifs, ST15, ST15reversion-inducing cysteine-rich protein with Kazal motifs, suppression of tumorigenicity 15 (reversion-inducing-cysteine-rich protein withkazal motifs), suppression of tumorigenicity 5 (reversion-inducing-cysteine-rich protein withkazal motifs), Suppressor of tumorigenicity 15 protein
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human RECK protein

Human RECK (Met1-Pro941) Accession # O95980IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in water.
Buffer Lyophilized from a 0.2 μm filtered solution in Tris and NaCl.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.