missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RND3 Protein

A cDNA sequence encoding the RND3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13328-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15913229

  • 212.00€ / 20µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RND3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MKERRASQKL SSKSIMDPNQ NVKCKIVVVG DSQCGKTALL HVFAKDCFPE NYVPTVFENY TASFEIDTQR IELSLWDTSG SPYYDNVRPL SYPDSDAVLI CFDISRPETL DSVLKKWKGE IQEFCPNTKM LLVGCKSDLR TDVSTLVELS NHRQTPVSYD QGANMAKQIG AATYIECSAL QSENSVRDIF HVATLACVNK TNKNVKRNKS QRATKRISHM PSRPELSAVA TDLRKDKAKS C
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RND3 Human solution containing 20mM Tris-HCl pH 8.0, 1mM DTT, 1mM EDTA, 0.1M NaCl and 40% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions