missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RNGTT Protein
A cDNA sequence encoding the RNGTT was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | RNGTT Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943239
|
enQuireBio™
QP13332-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923239
|
enQuireBio™
QP13332-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933239
|
enQuireBio™
QP13332-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RNGTT Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RNGTT protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5), 0.2M NaCl, 40% glycerol, 2mM DTT and 0.1mM PMSF. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAHNKIP PRWLNCPRRG QPVAGRFLPL KTMLGPRYDS QVAEENRFHP SMLSNYLKSL KVKMGLLVDL TNTSRFYDRN DIEKEGIKYI KLQCKGHGEC PTTENTETFI RLCERFNERN PPELIGVHCT HGFNRTGFLI CAFLVEKMDW SIEAAVATFA QARPPGIYKG DYLKELFRRY GDIEEAPPPP LLPDWCFEDD EDEDEDEDGK KESEPGSSAS FGKRRKERLK LGAIFLEGVT VKGVTQVTTQ PKLGEVQQKC HQFCGWEGSG FPGAQPVSMD KQNIKLLDLK PYKVSWKADG TRYMMLIDGT NEVFMIDRDN SVFHVSNLEF PFRKDLRMHL SNTLLDGEMI IDRVNGQAVP RYLIYDIIKF NSQPVGDCDF NVRLQCIERE IISPRHEKMK TGLIDKTQEP FSVRNKPFFD ICTSRKLLEG NFAKEVSHEM DGLIFQPTGK YKPGRCDDIL KWKPPSLNSV DFRLKITRMG GEGLLPQNVG LLYVGGYERP FAQIKVTKEL KQYDNKIIEC KFENNSWVFM RQRTDKSFPN AYNTAMAVCN SISNPVTKEM LFEFIDRCTA ASQGQKRKHH LDPDTELMPP PPPKRPRPLT | |
Greater than 85.0% as determined by SDS-PAGE. |