missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RNMTL1 Protein

Product Code. 15923249
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15923249

Brand: enQuireBio™ QP1333520ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the RNMTL1 was constructed and used to recombinantly synthesize the protein.

Specifications

Name RNMTL1 Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMAALVRP ARFVVRPLLQ VVQAWDLDAR RWVRALRRSP VKVVFPSGEV VEQKRAPGKQ PRKAPSEASA QEQREKQPLE ESASRAPSTW EESGLRYDKA YPGDRRLSSV MTIVKSRPFR EKQGKILLEG RRLISDALKA GAVPKMFFFS RLEYLKELPV DKLKGVSLIK VKFEDIKDWS DLVTPQGIMG IFAKPDHVKM TYPKTQLQHS LPLLLICDNL RDPGNLGTIL RSAAGAGCSK VLLTKGCVDA WEPKVLRAGM GAHFRMPIIN NLEWETVPNY LPPDTRVYVA DNCGLYAQAE MSNKASDHGW VCDQRVMKFH KYEEEEDVET GASQDWLPHV EVQSYDSDWT EAPAAVVIGG ETYGVSLESL QLAESTGGKR LLIPVVPGVD SLNSAMAASI LLFEGKRQLR GRAEDLSRDR SYH
Buffer RNMTL1 protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
Purity or Quality Grade Greater than 85.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.