missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RWDD4 Protein
A cDNA sequence encoding the RWDD4 was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | RWDD4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15923379
|
enQuireBio™
QP13377-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15903379
|
enQuireBio™
QP13377-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15913379
|
enQuireBio™
QP13377-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RWDD4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RWDD4 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSANEDQ EMELEALRSI YEGDESFREL SPVSFQYRIG ENGDPKAFLI EISWTETYPQ TPPILSMNAF FNNTISSAVK QSILAKLQEA VEANLGTAMT YTLFEYAKDN KEQFMENHNP INSATSISNI ISIETPNTAP SSKKKDKKEQ LSKAQKRKLA DKTDHKGELP RGWNWVDVVK HLSKTGSKDD E | |
Greater than 90.0% as determined by SDS-PAGE. |