missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SCGB3A2 Protein
A cDNA sequence encoding the SCGB3A2 was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | SCGB3A2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933539
|
enQuireBio™
QP13429-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15913539
|
enQuireBio™
QP13429-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923539
|
enQuireBio™
QP13429-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SCGB3A2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SCGB3A2 solution at a concentration of 0.4 mg/ml in phosphate buffered saline (PBS) pH 8.0. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MFLINKVPLP VDKLAPLPLD NILPFMDPLK LLLKTLGISV EHLVEGLRKC VNELGPEASE AVKKLLEALS HLVGGSGGSF LINKVPLPVD KLAPLPLDNI LPFMDPLKLL LKTLGISVEH LVEGLRKCVN ELGPEASEAV KKLLEALSHL VHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |