missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SCO1 Protein

A cDNA sequence encoding the SCO1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13435-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933559

  • 189.00€ / 20µg

Estimated Shipment: 01-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

SCO1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SCO1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGKPLLGGPF SLTTHTGERK TDKDYLGQWL LIYFGFTHCP DVCPEELEKM IQVVDEIDSI TTLPDLTPLF ISIDPERDTK EAIANYVKEF SPKLVGLTGT REEVDQVARA YRVYYSPGPK DEDEDYIVDH TIIMYLIGPD GEFLDYFGQN KRKGEIAASI ATHMRPYRKK SLEHHHHHH
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions