missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SH2D1B Protein
A cDNA sequence encoding the SH2D1B was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | SH2D1B Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933729
|
enQuireBio™
QP13486-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15913729
|
enQuireBio™
QP13486-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923729
|
enQuireBio™
QP13486-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SH2D1B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SH2D1B protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPMDLPYYH GRLTKQDCET LLLKEGVDGN FLLRDSESIP GVLCLCVSFK NIVYTYRIFR EKHGYYRIQT AEGSPKQVFP SLKELISKFE KPNQGMVVHL LKPIKRTSPS LRWRGLKLEL ETFVNSNSDY VDVLPHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |