missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SLAMF6 Protein

A cDNA sequence encoding the SLAMF6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13519-HIS-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15993829

  • 212.00€ / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SLAMF6 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSQSSLTPL MVNGILGESV TLPLEFPAGE KVNFITWLFN ETSLAFIVPH ETKSPEIHVT NPKQGKRLNF TQSYSLQLSN LKMEDTGSYR AQISTKTSAK LSSYTLRILR QLRNIQVTNH SQLFQNMTCE LHLTCSVEDA DDNVSFRWEA LGNTLSSQPN LTVSWDPRIS SEQDYTCIAE NAVSNLSFSV SAQKLCEDVK IQYTDTKM
Greater than 85.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SLAMF6 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4 Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions