missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SNCB Protein

A cDNA sequence encoding the SNCB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13536-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933889

  • 125.00€ / 10µg

Estimated Shipment: 25-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

SNCB Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein (1 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.1M NaCl and 1mM MgCl2.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MDVFMKGLSM AKEGVVAAAE KTKQGVTEAA EKTKEGVLYV GSKTREGVVQ GVASVAEKTK EQASHLGGAV FSGAGNIAAA TGLVKREEFP TDLKPEEVAQ EAAEEPLIEP LMEPEGESYE DPPQEEYQEY EPEA
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions