missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNTA1 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | SNTA1 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMASGRRA PRTGLLELRA GAGSGAGGER WQRVLLSLAE DVLTVSPADG DPGPEPGAPR EQEPAQLNGA AEPGAGPPQL PEALLLQRRR VTVRKADAGG LGISIKGGRE NKMPILISKI FKGLAADQTE ALFVGDAILS VNGEDLSSAT HDEAVQVLKK TGKEVVLEVK YMKDVSPYFK NSTGGTSVGW DSPPASPLQR QPSSPGPTPR NFSEAKHMSL KMAYVSKRCT PNDPEPRYLE ICSADGQDTL FLRAKDEASA RSWATAIQAQ VNTLTPRVKD ELQALLAATS TAGSQDIKQI GWLTEQLPSG GTAPTLALLT EKELLLYLSL PETREALSRP ARTAPLIATR LVHSGPSKGS VPYDAELSFA LRTGTRHGVD THLFSVESPQ ELAAWTRQLV DGCHRAAEGV QEVSTACTWN GRPCSLSVHI DKGFTLWAAE PGAARAVLLR QPFEKLQMSS DDGASLLFLD FGGAEGEIQL DLHSCPKTIV FIIHSFLSAK VTRLGLLA |
| Buffer | SNTA1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.15M NaCl. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction