missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SOD2 Protein

Product Code. p-7148676
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Unit Size:
1mg
25µg
5µg
This item is not returnable. View return policy

Product Code. 15945408

Brand: enQuireBio™ QP108665ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the SOD2 was constructed and used to recombinantly synthesize the protein.

Specifications

Name SOD2 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Specific activity is > 1,200 units/mg, in which one unit will inhibit the rate of reduction of cytochrome c by 50% in a coupled system, using xanthine and Xanthine oxidase at pH 7.8 at 25C in a 1.5 ml reaction volume.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MKHSLPDLPY DYGALEPHIN AQIMQLHHSK HHAAYVNNLN VTEEKYQEAL AKGDVTAQIA LQPALKFNGG GHINHSIFWT NLSPNGGGEP KGELLEAIKR DFGSFDKFKE KLTAASVGVQ GSGWGWLGFN KERGHLQIAA CPNQDPLQGT TGLIPLLGID VWEHAYYLQY KNVRPDYLKA IWNVINWENV TERYMACKK
Buffer SOD2 Human solution containing 20mM Tris-HCl pH 8 and 20% glycerol.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.