missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SOD2 Protein
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Unit Size:
1mg
25µg
5µg
Specifications
Specifications
| Name | SOD2 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | Specific activity is > 1,200 units/mg, in which one unit will inhibit the rate of reduction of cytochrome c by 50% in a coupled system, using xanthine and Xanthine oxidase at pH 7.8 at 25C in a 1.5 ml reaction volume. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MKHSLPDLPY DYGALEPHIN AQIMQLHHSK HHAAYVNNLN VTEEKYQEAL AKGDVTAQIA LQPALKFNGG GHINHSIFWT NLSPNGGGEP KGELLEAIKR DFGSFDKFKE KLTAASVGVQ GSGWGWLGFN KERGHLQIAA CPNQDPLQGT TGLIPLLGID VWEHAYYLQY KNVRPDYLKA IWNVINWENV TERYMACKK |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction