missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SPR Protein

A cDNA sequence encoding the SPR was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13582-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924049

  • 3740.00€ / 1mg

Estimated Shipment: 28-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

SPR Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The SPR solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MEGGLGRAVC LLTGASRGFG RTLAPLLASL LSPGSVLVLS ARNDEALRQL EAELGAERSG LRVVRVPADL GAEAGLQQLL GALRELPRPK GLQRLLLINN AGSLGDVSKG FVDLSDSTQV NNYWALNLTS MLCLTSSVLK AFPDSPGLNR TVVNISSLCA LQPFKGWALY CAGKAARDML FQVLALEEPN VRVLNYAPGP LDTDMQQLAR ETSVDPDMRK GLQELKAKGK LVDCKVSAQK LLSLLEKDEF KSGAHVDFYD K
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions