missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SRA1 Protein
A cDNA sequence encoding the SRA1 was constructed and used to recombinantly synthesize the protein.
137.00€ - 1815.00€
Specifications
Gene ID (Entrez) | 10011 |
---|---|
Name | SRA1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | SRA1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954069
|
enQuireBio™
QP13589-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944069
|
enQuireBio™
QP13589-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934069
|
enQuireBio™
QP13589-100UG |
100 μg |
1815.00€
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10011 | |
Research Use Only | |
SRA1 | |
Human | |
His | |
SRA1 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) , 0.1MNaCl ,10% glycerol and 1mM DTT. |
SRA1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSVGSGPAS GVEPTSFPVE SEAVMEDVLR PLEQALEDCR GHTRKQVCDD ISRRLALLQE QWAGGKLSIP VKKRMALLVQ ELSSHRWDAA DDIHRSLMVD HVTEVSQWMV GVKRLIAEKR SLFSEEAANE EKSAATAEKN HTIPGFQQAS | |
Greater than 85.0% as determined by SDS-PAGE. |