missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STC 2 Protein

A cDNA sequence encoding the STC 2 was constructed and used to recombinantly synthesize the protein.

137.00€ - 4090.00€

Specifications

Gene ID (Entrez) 8614
Name STC 2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol STC2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15934149
View Documents Promotion Details
enQuireBio™
QP13613-HIS-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15924149
View Documents Promotion Details
enQuireBio™
QP13613-HIS-20UG
20 μg
212.00€
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15914149
View Documents Promotion Details
enQuireBio™
QP13613-HIS-1MG
1 mg
4090.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

8614
Research Use Only
STC2
Human
His
Filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM Tris buffer, 20mM NaCl, pH 7.5.
STC 2 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
TDATNPPEGP QDRSSQQKGR LSLQNTAEIQ HCLVNAGDVG CGVFECFENN SCEIRGLHGI CMTFLHNAGKFDAQGKSFIK DALKCKAHAL RHRFGCISRK CPAIREMVSQ LQRECYLKHD LCAAAQENTR VIVEMIHFKDLLLHEPYVDL VNLLLTCGEE VKEAITHSVQ VQCEQNWGSL CSILSFCTSA IQKPPTAPPE RQPQVDRTKLSRAHHGEAGH HLPEPSSRET GRGAKGERGS KSHPNAHARG RVGGLGAQGP SGSSEWEDEQ SEYSDIRRAAADYKDDDDK
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications