missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Steroid 21-hydroxylase Protein

A DNA sequence encoding the Homo sapiens (Human) Steroid 21-hydroxylase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5907-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978579

  • 2385.00€ / 1mg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000491.4
Steroid 21-hydroxylase Protein
Research Use Only
CYP21A2
Recombinant Protein
E. coli
MLLLGLLLLP LLAGARLLWN WWKLRSLHLP PLAPGFLHLL QPDLPIYLLG LTQKFGPIYR LHLGLQDVVV LNSKRTIEEA MVKKWADFAG RPEPLTYKLV SKNYPDLSLG DYSLLWKAHK KLTRSALLLG IRDSMEPVVE QLTQEFCERM RAQPGTPVAI EEEFSLLTCS IICYLTFGDK IKDDNLMPAY YKCIQEVLKT WSHWSIQIVD VIPFLRFFPN PGLRRLKQAI EKRDHIVEMQ LRQHKESLVA GQWRDMMDYM LQGVAQPSME EGSGQLLEGH VHMAAVDLLI GGTETTANTL SWAVVFLLHH PEIQQRLQEE LDHELGPGAS SSRVPYKDRA RLPLLNATIA EVLRLRPVVP LALPHRTTRP SSISGYDIPE GTVIIPNLQG AHLDETVWER PHEFWPDRFL EPGKNSRALA FGCGARVCLG EPLARLELFV VLTRLLQAFT LLPSGDALPS LQPLPHCSVI LKMQPFQVRL QPRGMGAHSP GQNQ
Greater than 80% as determined by SDS-PAGE.
1589
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions