missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Steroid 21-hydroxylase Protein
A DNA sequence encoding the Homo sapiens (Human) Steroid 21-hydroxylase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5907-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000491.4 | |
Steroid 21-hydroxylase Protein | |
Research Use Only | |
CYP21A2 | |
Recombinant Protein | |
E. coli | |
MLLLGLLLLP LLAGARLLWN WWKLRSLHLP PLAPGFLHLL QPDLPIYLLG LTQKFGPIYR LHLGLQDVVV LNSKRTIEEA MVKKWADFAG RPEPLTYKLV SKNYPDLSLG DYSLLWKAHK KLTRSALLLG IRDSMEPVVE QLTQEFCERM RAQPGTPVAI EEEFSLLTCS IICYLTFGDK IKDDNLMPAY YKCIQEVLKT WSHWSIQIVD VIPFLRFFPN PGLRRLKQAI EKRDHIVEMQ LRQHKESLVA GQWRDMMDYM LQGVAQPSME EGSGQLLEGH VHMAAVDLLI GGTETTANTL SWAVVFLLHH PEIQQRLQEE LDHELGPGAS SSRVPYKDRA RLPLLNATIA EVLRLRPVVP LALPHRTTRP SSISGYDIPE GTVIIPNLQG AHLDETVWER PHEFWPDRFL EPGKNSRALA FGCGARVCLG EPLARLELFV VLTRLLQAFT LLPSGDALPS LQPLPHCSVI LKMQPFQVRL QPRGMGAHSP GQNQ | |
Greater than 80% as determined by SDS-PAGE. |
1589 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |