missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STOML1 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | STOML1 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSLKIVPTY ERMIVFRLGR IRTPQGPGMV LLLPFIDSFQ RVDLRTRAFN VPPCKLASKD GAVLSVGADV QFRIWDPVLS VMTVKDLNTA TRMTAQNAMT KALLKRPLRE IQMEKLKISD QLLLEINDVT RAWGLEVDRV ELAVEAVLQP PQDSPAGPNL DSTLQQLALH FLGGSMNSMA GGAPSPGPAD TVEMVSEVEP PAPQVGARSS PKQPLAEGLL TALQPFLSEA LVSQVGACYQ FNVVLPSGTQ SAYFLDLTTG RGRVGHGVPD GIPDVVVEMA EADLRALLCR ELRPLGAYMS GRLKVKGDLA MAMKLEAVLR ALK |
| Buffer | STOML1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction