missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STX17 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | STX17 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMSEDEEK VKLRRLEPAI QKFIKIVIPT DLERLRKHQI NIEKYQRCRI WDKLHEEHIN AGRTVQQLRS NIREIEKLCL KVRKDDLVLL KRMIDPVKEE ASAATAEFLQ LHLESVEELK KQFNDEETLL QPPLTRSMTV GGAFHTTEAE ASSQSLTQIY ALPEIPQDQN AAESWETLEA DLIELSQLVT DFSLLVNSQQ EKIDSIADHV NSAAVNVEEG TKNLGKAAKY KL |
| Buffer | The STX17 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 30% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction