missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SUMO2 Protein

Product Code. 15904269
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 15904269

Brand: enQuireBio™ QP1364910ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the SUMO2 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 6613
Name SUMO2 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol SUMO2
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MADEKPKEGV KTENNDHINL KVAGQDGSVV QFKIKRHTPL SKLMKAYCER QGLSMRQIRF RFDGQPINET DTPAQLEMED EDTIDVFQQQ TGGVY
Buffer SUMO2 at 100microg/ml in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.