missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TGF-beta 3 Protein
Click to view available options
Quantity:
10 μg
2 μg
200 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Unit Size:
10µg
200µg
2µg
Description
Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED50 for this effect is 0.01-0.04 ng/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
| Gene ID (Entrez) | 7043 |
| Molecular Weight (g/mol) | 12.7 kDa (monomer) |
| Name | TGF-beta 3 |
| Quantity | 2 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human TGF-beta 3 protein Ala301-Ser412 (Tyr340Phe) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction