missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human THAP1 Protein

A cDNA sequence encoding the THAP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13739-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924549

  • 189.00€ / 20µg

Estimated Shipment: 28-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

THAP1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
THAP1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2M Urea, 10% glycerol, 1mM DTT and 0.2M NaCl.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMVQSCS AYGCKNRYDK DKPVSFHKFP LTRPSLCKEW EAAVRRKNFK PTKYSSICSE HFTPDCFKRE CNNKLLKENA VPTIFLCTEP HDKKEDLLEP QEQLPPPPLP PPVSQVDAAI GLLMPPLQTP VNLSVFCDHN YTVEDTMHQR KRIHQLEQQV EKLRKKLKTA QQRCRRQERQ LEKLKEVVHF QKEKDDVSER GYVILPNDYF EIVEVPA
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions