missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thioredoxin-interacting Protein

A DNA sequence encoding the Homo sapiens(Human) Thioredoxin-interacting protein, was expressed in the hosts and tags indicated.

469.00€ - 1705.00€

Specifications

Accession Number NP_001300901.1
Gene ID (Entrez) 10628
Name Thioredoxin-interacting Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15932768
View Documents Promotion Details
enQuireBio™
QP10023-IV-100UG
100 μg
1705.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15952768
View Documents Promotion Details
enQuireBio™
QP10023-IV-50UG
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15942768
View Documents Promotion Details
enQuireBio™
QP10023-IV-10UG
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001300901.1
Thioredoxin-interacting Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
10628
Research Use Only
Txnip
Recombinant Protein
E. coli
MVMFKKIKSF EVVFNDPEKV YGSGEKVAGR VIVEVCEVTR VKAVRILACG VAKVLWMQGS QQCKQTSEYL RYEDTLLLED QPTGENEMVI MRPGNKYEYK FGFELPQGPL GTSFKGKYGC VDYWVKAFLD RPSQPTQETK KNFEVVDLVD VNTPDLMAPV SAKKEKKVSC MFIPDGRVSV SARIDRKGFC EGDEISIHAD FENTCSRIVV PKAAIVARHT YLANGQTKVL TQKLSSVRGN HIISGTCASW RGKSLRVQKI RPSILGCNIL RVEYSLLIYV SVPGSKKVIL DLPLVIGSRS GLSSRTSSMA SRTSSEMSWV DLNIPDTPEA PPCYMDVIPE DHRLESPTTP LLDDMDGSQD SPIFMYAPEF KFMPPPTYTE VDPCILNNNV Q
Greater than 85% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications