missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Thrombopoietin/TPO GMP Protein, CF

Product Code. 30306108
Click to view available options
Quantity:
1 mg
50 μg
Unit Size:
1mg
50µg
This item is not returnable. View return policy

Product Code. 30306108

Brand: R&D Systems™ BTTPOGMP01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. Avanzi, G. et al. (1988) Br. J. Haematol. 69:359. The ED50 for this effect is 0.0500-0.500 ng/mL. The specific activity of Recombinant Human Thrombopoietin is >1.00 x 107 units/mg, which is calibrated against the Human Thrombopoietin reference standard (NIBSC code: 03/124).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_000451.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 7066
Molecular Weight (g/mol) M.W.-theoretical: 18.7 kDa, M.W.-observed: 18-19 kDa, under reducing conditions.
Quantity 1 mg
Source E. coli-derived human Thrombopoietin/Tpo protein Ser22 - Leu195 with an N-terminal Alanine Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidelines.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 12 months when stored at ≤ -20 °C as supplied. Refer to lot specific COA for the Use by Date. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, ≤ -20 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in sterile water.
Purity or Quality Grade >95%, by SDS-PAGE with quantitative densitometry by Coomassie™ Blue Staining. 18732 Da ± 50 Da
Protein Thrombopoietin/THPO
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.