missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Thrombopoietin/TPO GMP Protein, CF
Click to view available options
Quantity:
1 mg
50 μg
Unit Size:
1mg
50µg
Description
Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. Avanzi, G. et al. (1988) Br. J. Haematol. 69:359. The ED50 for this effect is 0.0500-0.500 ng/mL. The specific activity of Recombinant Human Thrombopoietin is >1.00 x 107 units/mg, which is calibrated against the Human Thrombopoietin reference standard (NIBSC code: 03/124).
Specifications
Specifications
| Accession Number | NP_000451.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose. |
| Gene ID (Entrez) | 7066 |
| Molecular Weight (g/mol) | M.W.-theoretical: 18.7 kDa, M.W.-observed: 18-19 kDa, under reducing conditions. |
| Quantity | 50 μg |
| Source | E. coli-derived human Thrombopoietin/Tpo protein Ser22 - Leu195 with an N-terminal Alanine Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidelines. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 12 months when stored at ≤ -20 °C as supplied. Refer to lot specific COA for the Use by Date. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, ≤ -20 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction