missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIAL1 Protein
A cDNA sequence encoding the TIAL1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13749-5ug
Additional Details : Weight : 0.01000kg
Specifications
TIAL1 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TIAL1 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 50% glycerol, 2mM DTT and 1mM EDTA . |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMMEDDGQ PRTLYVGNLS RDVTEVLILQ LFSQIGPCKS CKMITEHTSN DPYCFVEFYE HRDAAAALAA MNGRKILGKE VKVNWATTPS SQKKDTSNHF HVFVGDLSPE ITTEDIKSAF APFGKISDAR VVKDMATGKS KGYGFVSFYN KLDAENAIVH MGGQWLGGRQ IRTNWATRKP PAPKSTQENN TKQLRFEDVV NQSSPKNCTV YCGGIASGLT DQLMRQTFSP FGQIMEIRVF PEKGYSFVRF STHESAAHAI VSVNGTTIEG HVVKCYWGKE SPDMTKNFQQ VDYSQWGQWS QVYGNPQQYG QYMANGWQVP PYGVYGQPWN QQGFGVDQSP SAAWMGGFGA QPPQGQAPPP VIPPPNQAGY GMASYQTQ |