missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNFRSF10D Protein

A cDNA sequence encoding the TNFRSF10D was constructed and used to recombinantly synthesize the protein.

137.00€ - 1815.00€

Specifications

Name TNFRSF10D Protein
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Measured in a neutralizing assay using Jurkat human T lymphocyte. The ED50 for this effect is less or equal to 10 ng/ml in the presence of 2ng/ml TRAIL.
Product Type Recombinant Protein
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15915478
View Documents Promotion Details
enQuireBio™
QP10888-1UG
1 μg
137.00€
1µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15935478
View Documents Promotion Details
enQuireBio™
QP10888-5UG
5 μg
212.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15925478
View Documents Promotion Details
enQuireBio™
QP10888-50UG
50 μg
1815.00€
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

TNFRSF10D Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Baculovirus
ATIPRQDEVP QQTVAPQQQR RSLKEEECPA GSHRSEYTGA CNPCTEGVDY TIASNNLPSC LLCTVCKSGQ TNKSSCTTTR DTVCQCEKGS FQDKNSPEMC RTCRTGCPRG MVKVSNCTPR SDIKCKNESA ASSTGKTPAA EETVTTILGM LASPYHVEPK SCDKTHTCPP CPAPELLGGPSVFLFPPKPK DTLMISRTPE VTCVVVDVSH EDPEVKFNWY VDGVEVHNAK TKPREEQYNS TYRVVSVLTV LHQDWLNGKE YKCKVSNKAL PAPIEKTISK AKGQPREPQV YTLPPSRDEL TKNQVSLTCL VKGFYPSDIA VEWESNGQPE NNYKTTPPVL DSDGSFFLYS KLTVDKSRWQQGNVFSCSVM HEALHNHYTQ KSLSLSPGKH HHHHH
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Measured in a neutralizing assay using Jurkat human T lymphocyte. The ED50 for this effect is less or equal to 10 ng/ml in the presence of 2ng/ml TRAIL.
Human
His
TNFRSF10D protein solution (0.5 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol.
Videos
SDS
Documents

Documents

Product Certifications