missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRIM33 Protein
A cDNA sequence encoding the TRIM33 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13816-2ug
Additional Details : Weight : 0.01000kg
Specifications
TRIM33 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TRIM33 is supplied in 20mM HEPES buffer pH 7.9, 550mM NaCl and 6M Urea. |
Greater than 80.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
MAENKGGGEA ESGGGGSGSA PVTAGAAGPA AQEAEPPLTA VLVEEEEEEG GRAGAEGGAA GPDDGGVAAA SSGSAQAASS PAASVGTGVA GGAVSTPAPA PASAPAPGPS AGPPPGPPAS LLDTCAVCQQ SLQSRREAEP KLLPCLHSFC LRCLPEPERQ LSVPIPGGSN GDIQQVGVIR CPVCRQECRQ IDLVDNYFVK DTSEAPSSSD EKSEQVCTSC EDNASAVGFC VECGEWLCKT CIEAHQRVKF TKDHLIRKKE DVSESVGASG QRPVFCPVHK QEQLKLFCET CDRLTCRDCQ LLEHKEHRYQ FLEEAFQNQK GAIENLLAKL LEKKNYVHFA ATQVQNRIKE VNETNKRVEQ EIKVAIFTLI NEINKKGKSL LQQLENVTKE RQMKLLQQQN DITGLSRQVK HVMNFTNWAI ASGSSTALLY SKRLITFQLR HILKARCDPV PAANGAIRFH CDPTFWAKNV VNLGNLVIES KPAPGYTPNV VVGQVPPGTN HISKTPGQIN LAQLRLQHMQ QQVYAQKHQQ LQQMRMQQPP APVPTTTTTT QQHPRQAAPQ MLQQQPPRLI SVQTMQRGNM NCGAFQAHQM RLAQNAARIP GIPRHSGPQY SMMQPHLQRQ HSNPGHAGPF PVVSVHNTTI NPTSPTTATM ANANRGPTSP SVTAIELIPS VTNPENLPSL PDIPPIQLED AGSSSLDNLL SRYISGSHLP PQPTSTMNPS PGPSALSPGS SGLSNSHTPV RPPSTSSTGS RGSCGSSGRT AEKTSLSFKS DQVKVKQEPG TEDEICSFSG GVKQEKTEDG RRSACMLSSP ESSLTPPLST NLHLESELDA LASLENHVKI EPADMNESCK QSGLSSLVNG KSPIRSLMHR SARIGGDGNN KDDDPNEDWC AVCQNGGDLL CCEKCPKVFH LTCHVPTLLS FPSGDWICTF CRDIGKPEVE YDCDNLQHSK KGKTAQGLSP VDQRKCERLL LYLYCHELSI EFQEPVPASI PNYYKIIKKP MDLSTVKKKL QKKHSQHYQI PDDFVADVRL IFKNCERFNE MMKVVQVYAD TQEINLKADS EVAQAGKAVA LYFEDKLTEI YSDRTFAPLP EFEQEEDDGE VTEDSDEDFI QPRRKRLKSD ERPVHIK |