missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tripartite motif-containing protein 72 Protein
A DNA sequence encoding the Homo sapiens (Human) Tripartite motif-containing protein 72, was expressed in the hosts and tags indicated.
605.00€ - 1690.00€
Specifications
Accession Number | NP_001008275.2 |
---|---|
Gene ID (Entrez) | 493829 |
Name | Tripartite motif-containing protein 72 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15922888
|
enQuireBio™
QP10131-MA-20UG |
20 μg |
605.00€
20µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15912888
|
enQuireBio™
QP10131-MA-100UG |
100 μg |
1690.00€
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932888
|
enQuireBio™
QP10131-MA-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
NP_001008275.2 | |
Tripartite motif-containing protein 72 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
Flag | |
Tris-based buffer, 50% glycerol |
493829 | |
Research Use Only | |
TRIM72 | |
Recombinant Protein | |
Mammal | |
MSAAPGLLHQ ELSCPLCLQL FDAPVTAECG HSFCRACLGR VAGEPAADGT VLCPCCQAPT RPQALSTNLQ LARLVEGLAQ VPQGHCEEHL DPLSIYCEQD RALVCGVCAS LGSHRGHRLL PAAEAHARLK TQLPQQKLQL QEACMRKEKS VAVLEHQLVE VEETVRQFRG AVGEQLGKMR VFLAALEGSL DREAERVRGE AGVALRRELG SLNSYLEQLR QMEKVLEEVA DKPQTEFLMK YCLVTSRLQK ILAESPPPAR LDIQLPIIS | |
Greater than 90% as determined by SDS-PAGE. |