missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Trypsin-1 Protein

A cDNA sequence encoding the Trypsin-1 was constructed and used to recombinantly synthesize the protein.

137.00€ - 5450.00€

Specifications

Name Trypsin-1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity 1 unit hydrolyzes 1 micromole of Na-Benzoyl-D,L-arginine 4-nitroanilide (BANI) per minute at 37C, pH 8.0, in the presence of 10mM calcium. The specific activity is >1,000 Units per mg protein as measured under the described conditions.
Product Type Recombinant Protein
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15935508
View Documents Promotion Details
enQuireBio™
QP10898-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15925508
View Documents Promotion Details
enQuireBio™
QP10898-20UG
20 μg
212.00€
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15915508
View Documents Promotion Details
enQuireBio™
QP10898-1MG
1 mg
5450.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Trypsin-1 Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Human
IVGGYNCEEN SVPYQVSLNS GYHFCGGSLI NEQWVVSAGH CYKSRIQVRL GEHNIEVLEG NEQFINAAKI IRHPQYDRKT LNNDIMLIKL SSRAVINARV STISLPTAPP ATGTKCLISG WGNTASSGAD YPDELQCLDA PVLSQAKCEA SYPGKITSNM FCVGFLEGGK DSCQGDSGGP VVCNGQLQGV VSWGDGCAQK NKPGVYTKVY NYVKWIKNTI AANS
Greater than 95% as determined by SDS-PAGE.
Research Use Only
1 unit hydrolyzes 1 micromole of Na-Benzoyl-D,L-arginine 4-nitroanilide (BANI) per minute at 37C, pH 8.0, in the presence of 10mM calcium. The specific activity is >1,000 Units per mg protein as measured under the described conditions.
Human
Untagged
The Trypsin-1 filtered 0.22um solution contains 20mM TRIS-HCl buffer pH 7.2.
Videos
SDS
Documents

Documents

Product Certifications