missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBA5 Protein
A cDNA sequence encoding the UBA5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13851-2ug
Additional Details : Weight : 0.01000kg
Specifications
UBA5 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UBA5 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 50mM NaCl. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAESVE RLQQRVQELE RELAQERSLQ VPRSGDGGGG RVRIEKMSSE VVDSNPYSRL MALKRMGIVS DYEKIRTFAV AIVGVGGVGS VTAEMLTRCG IGKLLLFDYD KVELANMNRL FFQPHQAGLS KVQAAEHTLR NINPDVLFEV HNYNITTVENFQHFMDRISN GGLEEGKPVD LVLSCVDNFE ARMTINTACN ELGQTWMESG VSENAVSGHI QLIIPGESAC FACAPPLVVA ANIDEKTLKR EGVCAASLPT TMGVVAGILV QNVLKFLLNF GTVSFYLGYN AMQDFFPTMS MKPNPQCDDR NCRKQQEEYK KKVAALPKQE VIQEEEEIIH EDNEWGIELV SEVSEEELKN FSGPVPDLPE GITVAYTIPK KQEDSVTELT VEDSGESLED LMAKMKNM |