missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2C Protein
A cDNA sequence encoding the UBE2C was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13854-1mg
Additional Details : Weight : 0.01000kg
Specifications
UBE2C Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASQNRD PAATSVAAAR KGAEPSGGAA RGPVGKRLQQ ELMTLMMSGD KGISAFPESD NLFKWVGTIH GAAGTVYEDL RYKLSLEFPS GYPYNAPTVK FLTPCYHPNV DTQGNICLDI LKEKWSALYD VRTILLSIQS LLGEPNIDSP LNTHAAELWK NPTAFKKYLQ ETYSKQVTSQ EP | |
Greater than 90.0% as determined by Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UBE2C protein 1 mg/ml is supplied in 20mM Tris-HCL, pH 8.0, 0.15M NaCl, 1mM DTT and 10% Glycerol. |