missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UBE2C Protein

A cDNA sequence encoding the UBE2C was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13854-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15914919

  • 212.00€ / 20µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UBE2C Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMASQNRD PAATSVAAAR KGAEPSGGAA RGPVGKRLQQ ELMTLMMSGD KGISAFPESD NLFKWVGTIH GAAGTVYEDL RYKLSLEFPS GYPYNAPTVK FLTPCYHPNV DTQGNICLDI LKEKWSALYD VRTILLSIQS LLGEPNIDSP LNTHAAELWK NPTAFKKYLQ ETYSKQVTSQ EP
Greater than 90.0% as determined by Analysis by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
UBE2C protein 1 mg/ml is supplied in 20mM Tris-HCL, pH 8.0, 0.15M NaCl, 1mM DTT and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions