missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2H Protein
A cDNA sequence encoding the UBE2H was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Name | UBE2H Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934939
|
enQuireBio™
QP13861-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924939
|
enQuireBio™
QP13861-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914939
|
enQuireBio™
QP13861-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
UBE2H Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UBE2H solution (1 mg/ml) contains 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 1mM DTT and 50mM NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSSPSPG KRRMDTDVVK LIESKHEVTI LGGLNEFVVK FYGPQGTPYE GGVWKVRVDL PDKYPFKSPS IGFMNKIFHP NIDEASGTVC LDVINQTWTA LYDLTNIFES FLPQLLAYPN PIDPLNGDAA AMYLHRPEEY KQKIKEYIQK YATEEALKEQ EEGTGDSSSE SSMSDFSEDE AQDMEL | |
Greater than 95.0% as determined by SDS-PAGE. |