missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ULBP4 Protein

A cDNA sequence encoding the ULBP4 was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Name ULBP4 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15935029
View Documents Promotion Details
enQuireBio™
QP13888-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15915029
View Documents Promotion Details
enQuireBio™
QP13888-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15925029
View Documents Promotion Details
enQuireBio™
QP13888-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

ULBP4 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ULBP4 was filtered (0.4 microm) and lyophilized in 0.05M Acetate buffer pH 4.0, 1% (w/v) Sucrose and 4% (w/v) Mannitol.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASHSLCFNFTIK SLSRPGQPWC EAQVFLNKNL FLQYNSDNNM VKPLGLLGKK VYATSTWGEL TQTLGEVGRD LRMLLCDIKP QIKTSDPSTL QVEMFCQREA ERCTGASWQF ATNGEKSLLF DAMNMTWTVI NHEASKIKET WKKDRGLEKY FRKLSKGDCD HWLREFLGHW EAMPEPTGRR ST
Greater than 85.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications