missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UROD Protein
A cDNA sequence encoding the UROD was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13907-25ug
Additional Details : Weight : 0.01000kg
Specifications
7389 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
UROD | |
Human | |
His | |
UROD Human solution containing 20mM Tris pH 8.0, 1mM DTT, 0.1M NaCl, 1mM EDTA and 20% glycerol. |
UROD Protein | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEANGLGPQG FPELKNDTFL RAAWGEETDY TPVWCMRQAG RYLPEFRETR AAQDFFSTCR SPEACCELTL QPLRRFPLDA AIIFSDILVV PQALGMEVTM VPGKGPSFPE PLREEQDLER LRDPEVVASE LGYVFQAITL TRQRLAGRVP LIGFAGAPWT LMTYMVEGGG SSTMAQAKRW LYQRPQASHQ LLRILTDALV PYLVGQVVAG AQALQLFESH AGHLGPQLFN KFALPYIRDV AKQVKARLRE AGLAPVPMII FAKDGHFALE ELAQAGYEVV GLDWTVAPKK ARECVGKTVT LQVNLDPCAL YASEEEIGQL VKQMLDDFGP HRYIANLGHG LYPDMDPEHV GAFVDAVHKH SRLLRQN |