missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VRK3 Protein

A cDNA sequence encoding the VRK3 was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Name VRK3 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15935209
View Documents Promotion Details
enQuireBio™
QP13947-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15915209
View Documents Promotion Details
enQuireBio™
QP13947-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15925209
View Documents Promotion Details
enQuireBio™
QP13947-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

VRK3 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The VRK3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 40% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMISFCPD CGKSIQAAFK FCPYCGNSLP VEEHVGSQTF VNPHVSSFQG SKRGLNSSFE TSPKKVKWSS TVTSPRLSLF SDGDSSESED TLSSSERSKG SGSRPPTPKS SPQKTRKSPQ VTRGSPQKTS CSPQKTRQSP QTLKRSRVTT SLEALPTGTV LTDKSGRQWK LKSFQTRDNQ GILYEAAPTS TLTCDSGPQK QKFSLKLDAK DGRLFNEQNF FQRAAKPLQV NKWKKLYSTP LLAIPTCMGF GVHQDKYRFL VLPSLGRSLQ SALDVSPKHV LSERSVLQVA CRLLDALEFL HENEYVHGNV TAENIFVDPE DQSQVTLAGY GFAFRYCPSG KHVAYVEGSR SPHEGDLEFI SMDLHKGCGP SRRSDLQSLG YCMLKWLYGF LPWTNCLPNT EDIMKQKQKF VDKPGPFVGP CGHWIRPSET LQKYLKVVMA LTYEEKPPYA MLRNNLEALL QDLRVSPYDP IGLPMVP
Greater than 85% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications