missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VRK3 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | VRK3 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMISFCPD CGKSIQAAFK FCPYCGNSLP VEEHVGSQTF VNPHVSSFQG SKRGLNSSFE TSPKKVKWSS TVTSPRLSLF SDGDSSESED TLSSSERSKG SGSRPPTPKS SPQKTRKSPQ VTRGSPQKTS CSPQKTRQSP QTLKRSRVTT SLEALPTGTV LTDKSGRQWK LKSFQTRDNQ GILYEAAPTS TLTCDSGPQK QKFSLKLDAK DGRLFNEQNF FQRAAKPLQV NKWKKLYSTP LLAIPTCMGF GVHQDKYRFL VLPSLGRSLQ SALDVSPKHV LSERSVLQVA CRLLDALEFL HENEYVHGNV TAENIFVDPE DQSQVTLAGY GFAFRYCPSG KHVAYVEGSR SPHEGDLEFI SMDLHKGCGP SRRSDLQSLG YCMLKWLYGF LPWTNCLPNT EDIMKQKQKF VDKPGPFVGP CGHWIRPSET LQKYLKVVMA LTYEEKPPYA MLRNNLEALL QDLRVSPYDP IGLPMVP |
| Buffer | The VRK3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 40% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction