missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WWOX Protein
A cDNA sequence encoding the WWOX was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | WWOX Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935269
|
enQuireBio™
QP13967-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915269
|
enQuireBio™
QP13967-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925269
|
enQuireBio™
QP13967-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
WWOX Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The WWOX solution (1 mg/ml) contains 20mM Tris pH 8.0, and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAALRYAGLD DTDSEDELPP GWEERTTKDG WVYYANHTEE KTQWEHPKTG KRKRVAGDLP YGWEQETDEN GQVFFVDHIN KRTTYLDPRL AFTVDDNPTK PTTRQRYDGS TTAMEILQGR DFTGKVVVVT GANSGIGFET AKSFALHGAH VILACRNMAR ASEAVSRILE EWQQGAATTV YCAAVPELEG LGGMYFNNCC RCMPSPEAQS EETARTLWAL SERLIQERLG SQSG | |
Greater than 85.0% as determined by SDS-PAGE. |