missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human WWOX Protein

A cDNA sequence encoding the WWOX was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13967-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15925269

  • 7200.00€ / 1mg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

WWOX Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The WWOX solution (1 mg/ml) contains 20mM Tris pH 8.0, and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAALRYAGLD DTDSEDELPP GWEERTTKDG WVYYANHTEE KTQWEHPKTG KRKRVAGDLP YGWEQETDEN GQVFFVDHIN KRTTYLDPRL AFTVDDNPTK PTTRQRYDGS TTAMEILQGR DFTGKVVVVT GANSGIGFET AKSFALHGAH VILACRNMAR ASEAVSRILE EWQQGAATTV YCAAVPELEG LGGMYFNNCC RCMPSPEAQS EETARTLWAL SERLIQERLG SQSG
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions