missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZCCHC17 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | ZCCHC17 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMNSGRPE TMENLPALYT IFQGEVAMVT DYGAFIKIPG CRKQGLVHRT HMSSCRVDKP SEIVDVGDKV WVKLIGREMK NDRIKVSLSM KVVNQGTGKD LDPNNVIIEQ EERRRRSFQD YTGQKITLEA VLNTTCKKCG CKGHFAKDCF MQPGGTKYSL IPDEEEEKEE AKSAEFEKPD PTRNPSRKRK KEKKKKKHRD RKSSDSDSSD SESDTGKRAR HTSKDSKAAK KKKKKKKHKK KHKE |
| Buffer | The ZCCHC17 protein solution (0.25 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 2mM DTT, 0.1mM PMSF and 20% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction