missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Tokio Protein
A cDNA sequence encoding the Influenza-B Tokio was constructed and used to recombinantly synthesize the protein.
137.00€ - 3030.00€
Specifications
Name | Influenza-B Tokio Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15910439
|
enQuireBio™
QP12443-10UG |
10 μg |
137.00€
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930439
|
enQuireBio™
QP12443-50UG |
50 μg |
212.00€
50µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920439
|
enQuireBio™
QP12443-1MG |
1 mg |
3030.00€
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Especificaciones
Influenza-B Tokio Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Influenza B virus | |
The B/Tokio/53/99solution (1.7 mg/ml) contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal. |