missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H1N1 Puerto Rico Recombinant Protein
A cDNA sequence encoding the H1N1 Puerto Rico Recombinant was constructed and used to recombinantly synthesize the protein.
137.00€ - 2725.00€
Specifications
Name | Influenza H1N1 Puerto Rico Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929298
|
enQuireBio™
QP12077-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919298
|
enQuireBio™
QP12077-10UG |
10 μg |
301.00€
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15909298
|
enQuireBio™
QP12077-100UG |
100 μg |
2725.00€
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H1N1 Puerto Rico Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant H1N1 A/Puerto Rico 08/1934 solution contains 10mM Sodium phosphate, pH 7.2, 150mM NaCl and 0.005% Tween-20. |