missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Panama Protein
Recombinant Protein
137.00€ - 3030.00€
Specifications
Name | Influenza H3N2 Panama Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969318
|
enQuireBio™
QP12085-10UG |
10 μg |
137.00€
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989318
|
enQuireBio™
QP12085-50UG |
50 μg |
212.00€
50µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979318
|
enQuireBio™
QP12085-1MG |
1 mg |
3030.00€
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H3N2 Panama Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The H3N2 A/Panama/2007/99 solution contains STE, 0.09% sodium azide (NaN3) and 0.005% thimerosal. |