missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H3N2 Wisconsin Protein

A cDNA sequence encoding the H3N2 Wisconsin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12091-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15969338

  • 137.00€ / 2µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza H3N2 Wisconsin Protein
Research Use Only
Recombinant Protein
Influenza H3N2
The Recombinant H3N2 A/Wisconsin/67/05 solution contains 10mM Sodium phosphate, pH 7.2 and 150mM Sodium Chloride.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions