missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse ACY1 Protein

A cDNA sequence encoding the ACY1 was constructed and used to recombinantly synthesize the protein.

137.00€ - 1815.00€

Specifications

Gene ID (Entrez) 109652
Name ACY1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol Acy1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15935688
View Documents Promotion Details
enQuireBio™
QP10957-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15925688
View Documents Promotion Details
enQuireBio™
QP10957-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15915688
View Documents Promotion Details
enQuireBio™
QP10957-100UG
100 μg
1815.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

109652
Research Use Only
Acy1
Mouse
His
ACY1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol.
ACY1 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSEFMTTKD PESEHPSVTL FRQYLRICTV QPNPDYGGAI TFLEERARQL GLSCQKIEVV PGFVITVLTW PGTNPSLPSI LLNSHTDVVP VFKEHWHHDP FEAFKDSEGY IYARGSQDMK SVSIQYLEAV RRLKSEGHRF PRTIHMTFVP DEEVGGHKGM ELFVKRPEFQ ALRAGFALDE GLANPTDAFT VFYSERSPWW VRVTSTGKPG HASRFIEDTA AEKLHKVISS ILAFREKERQ RLQANPHLKE GAVTSVNLTK LEGGVAYNVV PATMSASFDF RVAPDVDMKA FEKQLQRWCQ EAGEGVTFEF AQKFTEPRMT PTDDSDPWWA AFSGACKAMN LTLEPEIFPA ATDSRYIRAV GIPALGFSPM NRTPVLLHDH NERLHEDIFL RGVDIYTGLL SALASVPTLP GES
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications