missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Apolipoprotein C-III Protein

A DNA sequence encoding the Mus musculus (Mouse) Apolipoprotein C-III, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5667-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15926859

  • 3110.00€ / 1mg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001276685.1
Apolipoprotein C-III Protein
1 mg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
11814
Greater than 90% as determined by SDS-PAGE.
Research Use Only
APOC3
Recombinant Protein
E. coli
EEVEGSLLLG SVQGYMEQAS KTVQDALSSV QESDIAVVAR GWMDNHFRFL KGYWSKFTDK FTGFWDSNPE DQPTPAIES
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions