missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse beta-NGF / Beta-NGF Protein

Recombinant Protein

Brand:  enQuireBio™ QP5412-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936079

  • 112.00€ / 5µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_038637.1
beta-NGF / Beta-NGF Protein
Research Use Only
NGF
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
18049
5 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MSSTHPVFHM GEFSVCDSVS VWVGDKTTAT DIKGKEVTVL AEVNINNSVF RQYFFETKCR ASNPVESGCR GIDSKHWNSY CTTTHTFVKA LTTDEKQAAW RFIRIDTACV CVLSRKATRR G
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions