missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cathepsin K Protein
A DNA sequence encoding the Mus musculus (Mouse) Cathepsin K, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5890-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031828.2 | |
Cathepsin K Protein | |
Research Use Only | |
Ctsk | |
Recombinant Protein | |
E. coli | |
VPDSIDYRKK GYVTPVKNQG QCGSCWAFSS AGALEGQLKK KTGKLLALSP QNLVDCVTEN YGCGGGYMTT AFQYVQQNGG IDSEDAYPYV GQDESCMYNA TAKAAKCRGY REIPVGNEKA LKRAVARVGP ISVSIDASLA SFQFYSRGVY YDENCDRDNV NHAVLVVGYG TQKGSKHWII KNSWGESWGN KGYALLARNK NNACGITNMA SFPKM | |
Greater than 90% as determined by SDS-PAGE. |
13038 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |